Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

CPC® Metal Quick-Disconnect Fittings, Compression Inserts

Metal Quick-Disconnect Fitting, Compression Insert, Straight-Through, ¹/₄" OD, MC2004; 1/PK, Quick-Disconnect to Compression, Straight, Brass, chrome-plated, ⅛"
  MFLX31404-49
 :  MFLX31404-49
CPC® Metal Quick-Disconnect Fittings, Compression Inserts
  MFLX31404-49
CPC® Metal Quick-Disconnect Fittings, Compression Inserts
CPC® Metal Quick-Disconnect Fittings, Compression Inserts

Metal Quick-Disconnect Fitting, Compression Insert, Straight-Through, ¹/₄" OD, MC2004; 1/PK, Quick-Disconnect to Compression, Straight, Brass, chrome-plated, ⅛"

 
$357.00
   
Each (1UN)
 

Convenience of a quick-disconnect in pressure and vacuum applications.

  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Constructed of durable chrome-plated brass
  • Easy one-hand connection and disconnection

These CPC® metal quick-disconnect fittings are ideal for pressure and vacuum applications. Fittings are made of chrome-plated brass with stainless steel springs and latch. Valved fittings have an acetal shutoff valve that seals the flow path when fitting halves are disconnected. Can withstand vacuum to 250 psi, temperatures from -40 to 180 °F, and can be used with gases or liquids.

A complete coupler requires a fitting body and an insert with the same flow size (order separately).

 
  • Connection Type:
    Quick-Disconnect to Compression
  • Flow pattern:
    Straight
  • Description:
    Metal Quick-Disconnect Fitting, Compression Insert, Straight-Through, ¹/₄" OD, MC2004; 1/PK
  • Fitting Material:
    Brass, chrome-plated
  • Quick-disconnect Flow Size:
    ⅛"
  • Cat. No.:
    MFLX31404-49
  • Warranty:
    3 Months
  • Length:
    2.921 cm
  • Min. temperature:
    –40 °C (–40 °F)
  • Max. temperature:
    82 °C (180 °F)
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)
  • Max. operating pressure:
    250 psi
  • Seal material:
    Buna N
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.