Order Entry
Mexico
ContactUsLinkComponent
Fitting, Brass Compression Nut Assembly with Ferrule, 1/16" OD
Fitting, Brass Compression Nut Assembly with Ferrule, 1/16" OD
Catalog # MFLX31212-68
undefined
Fitting, Brass Compression Nut Assembly with Ferrule, 1/16" OD
Catalog # MFLX31212-68
Supplier Number:  IBD1FS+N
About this item
Can handle pressures up to 225 psi.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rated to 220 psi maximum at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).
  • Connection Type:
    Compression Nut Assembly with Ferrule
  • Flow pattern:
    Straight
  • Description:
    Fitting, Brass Compression Nut Assembly with Ferrule, ¹⁄₁₆" OD
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Cat. no.:
    MFLX31212-68
  • Fitting Material:
    Brass
Frequently Bought Together