Order Entry
Mexico
Orders LinkContactUsLinkComponent
PendoTECH® Single-Use Pressure Sensor Accessories

PendoTECH® Single-Use Pressure Sensor Accessories

Cable with RJ12 to Scilog Pump. 6 ft (1.8 m)
Catalog # MFLX19406-85
CE Compliant
Supplier:   PENDOTECH
Supplier #:  PDKT-650-298SC
PendoTECH® Single-Use Pressure Sensor Accessories
Catalog # MFLX19406-85
Supplier:  PENDOTECH
CE Compliant
PendoTECH® Single-Use Pressure Sensor Accessories
PendoTECH® Single-Use Pressure Sensor Accessories

Cable with RJ12 to Scilog Pump. 6 ft (1.8 m)

Price
$4,220.00
Log in for availability
Each (1UN)
Product Details & Documents

Optimize performance of your PendoTECH® Pressure Sensor.

  • Choose from a wide range of replacement parts and accessories

Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.

Documents

Specifications
  • Description:
    Cable with RJ12 to Scilog Pump. 6 ft (1.8 m)
  • Cat. No.:
    MFLX19406-85
  • Warranty:
    2 years
  • Length:
    182.88 cm (72")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.