PendoTECH® Single-Use Pressure Sensor Accessories
Reusable Temperature Sensor Cable with 1/4" Phone Jack for Hose Barb Fitting; 10 ft (3 m)
MFLX19406-15
CE Compliant
: PDKT-650-TEMPB
- Description:Reusable Temperature Sensor Cable with 1/4" Phone Jack for Hose Barb Fitting; 10 ft (3 m)
- Cat. No.:MFLX19406-15
- Warranty:2 years
- Length:304.8 cm (120")



