Order Entry
Mexico
ContactUsLinkComponent
Spirax Sarco Cast iron inverted bucket steam trap; 3/4" NPT(F), 30 psig
  MFLX07600-64
 :  Avantor
Spirax Sarco Cast iron inverted bucket steam trap; 3/4" NPT(F), 30 psig
  MFLX07600-64
 :  Avantor
Keep steam space free of condensate to provide maximum system efficiency.
  • All stainless steel working parts provide good resistance to corrosion and wear
  • Inverted bucket steam traps rely on the difference in density between steam and condensate to drain condensate as it forms.
  • Condensate is discharged close to steam temperature
Typical applications:
Steam main drip stations
Laundry equipment
Industrial dryers
Storage tanks
  • Connection Type:
    NPT(F)
  • Max. pressure:
    30 psi
  • Max. temperature:
    232 °C (450 °F)
  • Description:
    Cast iron inverted bucket steam trap; 3/4" NPT(F), 30 psig
  • Environmentally Preferable:
  • Wetted materials:
    Cast iron
  • Port size:
    0.75 NPT(F)
  • Warranty:
    1 year
  • Cat. no.:
    MFLX07600-64
  • Inlet Port:
    ¾" NPT(F)
  • Outlet Port:
    ¾" NPT(F)