Masterflex® Replacement Open-Head Sensor Shorting Plug for Sensor-Equipped Drives
MFLX07528-82
Make the most of your Masterflex® Open-Head Sensor with these accessories and replacement parts.
- Use in conjunction with Masterflex® L/S® and I/P® open-head sensor equipped pump heads
- Compatible with Masterflex® L/S® and I/P® pumps without integrated open-head sensor
- Controller accepts input signals from up to two pump heads
- Protect technicians and operators tasked with tubing changes and routine maintenance in busy production environments
- Controller is NEMA 4X / IP66 rated for washdown
The retrofit system is compatible with all I/P® pumps and with L/S® pumps that accept the Easy-Load® II pump head.
Use only with I/P® Easy-Load® pump head 77602-30 and with L/S® Easy-Load® II pump heads 77200-30 and 77200-32.
- Description:Replacement Masterflex Open-Head Sensor Shorting Plug for Sensor-Equipped Drives
- Environmentally Preferable:
- Warranty:2 years
- Cat. no.:MFLX07528-82
Frequently Bought Together