Masterflex® L/S® Tubing Loading Key for Standard Pump Heads
MFLX07013-90
Ensure proper tubing alignment and installation.
- Constructed of durable stainless steel
- Tubing key is suitable for use with L/S® standard pump heads
L/S® and I/P® Standard pump heads include tubing loading keys. A key is required to ensure proper tubing alignment and tension. Order a replacement key based on your pump head series, L/S® or I/P®.
- Description:tubing loading key for L/S Standard pump heads.
- Environmentally Preferable:
- Warranty:2 years
- Cat. no.:MFLX07013-90
- Pump Series:L/S
Frequently Bought Together