Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
L/S® Pump head mounting hardware, Standard and Easy-Load®, one Pump Head, Stainless Steel; 2/Set, 2 years warranty, Stainless steel, Masterflex L/S® Easy-Load® Pump head, L/S
Catalog # MFLX07013-04
Supplier: Avantor Fluid Handling
Supplier #: 7013-04
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
Catalog # MFLX07013-04
Supplier: Avantor Fluid Handling
Jump to:
Product Details & Documents
Easy stack multiple pump heads on your Masterflex® L/S® drive.
- Constructed of durable stainless steel
- See drive specifications for recommended number of heads
Stack multiple pump heads or replacement single pump head mounting hardware. Choose mounting hardware based on the Masterflex® L/S® pump head that you have. Compatible with the Masterflex L/S series pumps.
Specifications
- Description:L/S® Pump head mounting hardware, Standard and Easy-Load®, one Pump Head, Stainless Steel; 2/Set, 2 years warranty
- Material:Stainless steel
- Compatible with:Masterflex L/S® Easy-Load® Pump head
- Pump Series:L/S