Order Entry
Mexico
ContactUsLinkComponent
Tubing Rack, 5-Channel for 1/2" OD Tubing, Non-Adhesive Back, 10/Pk
Tubing Rack, 5-Channel for 1/2" OD Tubing, Non-Adhesive Back, 10/Pk
Catalog # MFLX06432-05
Supplier:  VWR International
undefined
Tubing Rack, 5-Channel for 1/2" OD Tubing, Non-Adhesive Back, 10/Pk
Catalog # MFLX06432-05
Supplier:  VWR International
About this item
An easy solution to secure and route multiple tubing lines.
  • Keep your tubing lines organized and untangled
These tube racks offer a simple and economical method for organizing tubing lines. The ³/₁₆" and ¹/₄" sizes can be easily separated down to single holders by hand for increased flexibility and custom tubing arrangements. The ³/₁₆" and ¹/₄" sizes are available with adhesive fastener, and all sizes are available with through holes for secure mounting (mounting screws are not included). Standard racks are black polyethylene; ³/₁₆" and ¹/₄" sizes are available in other colors.
  • Description:
    Tubing Rack, 5-Channel for 1/2" OD Tubing, Non-Adhesive Back, 10/Pk
  • Environmentally Preferable:
  • No. of channels:
    5
  • Warranty:
    1 year
  • Cat. no.:
    MFLX06432-05
  • Fits Tubing O.D.:
    12.7 mm (0.5")