Order Entry
Mexico
ContactUsLinkComponent
GF Signet Installation Tee Fitting, PVC 80, 1" Pipe ID
GF Signet Installation Tee Fitting, PVC 80, 1" Pipe ID
Catálogo # MFLX05620-23
undefined
GF Signet Installation Tee Fitting, PVC 80, 1" Pipe ID
Catálogo # MFLX05620-23
Supplier Number:  MPV8T010F

About this item

Easily install your flowmeters.

  • Constructed of durable CPVC/PVC 80

PVC Tee Fittings for PVC 80 pipes up to 4" in diameter and CPVC 80 pipes up to 1 1/2" in diameter. Slip on ends (no threads) sensors.

  • Description:
    Installation Tee Fitting. PVC 80. 1" Pipe ID
  • Environmentally Preferable:
  • Warranty:
    1 year part and 2 years labor
  • Connection type:
    1" ID Slip Fit
  • Cat. no.:
    MFLX05620-23
  • Fitting Material:
    PVC 80
Frequently Bought Together