Order Entry
Mexico
ContactUsLinkComponent
Masterflex® Tripod Base for Three to Five Tube Systems
Masterflex® Tripod Base for Three to Five Tube Systems
Catalog # MFLX03226-40
undefined
Masterflex® Tripod Base for Three to Five Tube Systems
Catalog # MFLX03226-40
Supplier Number:  TP3
About this item
Easily mount flowmeters on tripod for easy viewing.
  • Constructed of durable acrylic
These tripod bases are ideal for benchtop applications. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting. Tripod bases are made of acrylic and include a spirit leveler.
  • Description:
    Tripod Base for Three to Five Tube Systems
  • Environmentally Preferable:
  • Model:
    03226-40
  • Width:
    1755"
  • Warranty:
    1 year
  • Cat. no.:
    MFLX03226-40
  • Housing Material:
    Acrylic
Frequently Bought Together