$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
MSTRFLX TBNG PURIFLEX NO.17 3'
MFLX00437-CV
:
VWR International
MSTRFLX TBNG PURIFLEX NO.17 3'
MFLX00437-CV
:
VWR International
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Universal Pipette Tips
: VWR International