$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
P-DIOXANO RA ACS 4L CS4
85020-098
:
AVANTOR PERFORMANCE MATERIALS MX
P-DIOXANO RA ACS 4L CS4
85020-098
:
AVANTOR PERFORMANCE MATERIALS MX
:
M4937-08
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Universal Pipette Tips
: VWR International