Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
GUANTE ACCUTECH LATEX ESTERIL 6.5 CS200
Catalog # 85019-716
Proveedor:
ANSELL COMMERCIAL MEXICO SA DE MX
Certificados
Ampliar imagen
GUANTE ACCUTECH LATEX ESTERIL 6.5 CS200
Catalog # 85019-716
Proveedor:
ANSELL COMMERCIAL MEXICO SA DE MX
Supplier Number:
91-225/6.5
Certificados
Información relacionada
Documentation
ANSE91-225^6.5_2209503204.pdf
Documentation
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Proveedor: VWR International
VWR® Universal Pipette Tips
Proveedor: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
Proveedor: VWR International
VWR® Nitrile Examination Gloves, Blue
Proveedor: VWR International
VWR® Disposable Petri Dishes
Proveedor: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Proveedor: BDH