$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
VIAL MUESTRA 40ML TOC POR METH 4153 PK72
85019-662
:
REACTIVOS Y EQUIP SA DE CV MX
VIAL MUESTRA 40ML TOC POR METH 4153 PK72
85019-662
:
REACTIVOS Y EQUIP SA DE CV MX
:
HA2672700
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH