Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
JUEGO DE MANIJAS Y ABRAZADERAS
Catalog # 85019-658
Supplier:
BERKSHIRE CORPORATION MX
Expand image
JUEGO DE MANIJAS Y ABRAZADERAS
Catalog # 85019-658
Supplier:
BERKSHIRE CORPORATION MX
Supplier Number:
CHMH.SET.1
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Supplier: AIR-TITE
Sodium hydroxide 0.1 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
VWR® Self-Seal Sterilization Pouches
Supplier: VWR International
VWR® Razor Blades
Supplier: VWR International
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
VWR® Specimen Containers, Polypropylene with Polyethylene Caps
Supplier: VWR International