Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
HOSE TRANSFER SS CRYOGNC 12FT LN2 SYSTEM
Catalog # 76490-340
Supplier:
PHC CORPORATION OF NORTH AMERICA MX
Expand image
HOSE TRANSFER SS CRYOGNC 12FT LN2 SYSTEM
Catalog # 76490-340
Supplier:
PHC CORPORATION OF NORTH AMERICA MX
Supplier Number:
CVKLN2H12
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International