Objectives for REVOLVE Microscopes, Echo
Apochromat objective, 0.18 mm, 0.95 NA
Catalog # 76490-274
Supplier #: JMAG9402
Jump to:
Discontinued
Product Details & Documents
Documents
Echo Revolve
Product Literature
Specifications
- Magnification:40×
- Description:Apochromat objective
- Aperture:0.95 NA
- Working Distance:0.18 mm