$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
PM KIT BBD6220 / CYTOPERM
85019-406
:
THERMO FISHER SCIENTIFIC MX
PM KIT BBD6220 / CYTOPERM
85019-406
:
THERMO FISHER SCIENTIFIC MX
:
50076020
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH