$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
HYFLEX 11-840 LIGERO TAM 6 CS144
85019-028
:
ANSELL COMMERCIAL MEXICO SA DE MX
HYFLEX 11-840 LIGERO TAM 6 CS144
85019-028
:
ANSELL COMMERCIAL MEXICO SA DE MX
:
113036
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International