$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
PHOSPHATE PHOSPHORUS STD FOR IC 100ML
85018-554
:
SIGMA ALDRICH QUIMICA,S DE RL MX
PHOSPHATE PHOSPHORUS STD FOR IC 100ML
85018-554
:
SIGMA ALDRICH QUIMICA,S DE RL MX
:
95924-100ML
Frequently Bought Together
VWR® Disposable Serological Pipettes
: VWR International
VWR® Universal Pipette Tips
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International