$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
REFRIGERADOR TSG 25 SST SOLIDO
85017-880
:
THERMO FISHER SCIENTIFIC MX
REFRIGERADOR TSG 25 SST SOLIDO
85017-880
:
THERMO FISHER SCIENTIFIC MX
:
TSG25RSSA
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH