Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Screw Cap Microtubes, Secure Sample Storage

VWR® Screw Cap Microtubes, Secure Sample Storage

Microtubes without ribbing, Clear, 2.0 ml, Conical bottom, No, 20000×g, —, Non sterile
Catalog # 76417-250
Supplier:   VWR International
VWR® Screw Cap Microtubes, Secure Sample Storage
Catalog # 76417-250
Supplier:  VWR International
VWR® Screw Cap Microtubes, Secure Sample Storage
VWR® Screw Cap Microtubes, Secure Sample Storage

Microtubes without ribbing, Clear, 2.0 ml, Conical bottom, No, 20000×g, —, Non sterile

Price
$645.68
Log in for availability
Paquete de 500 Case de 10 (500UN)
Product Details & Documents

VWR® screw cap microtubes - secure storage for valuable samples. VWR® screw cap microtubes provide ultra-pure polypropylene construction, leak-proof caps, and sterile/non-sterile options for reliable lab sample storage.

  • Ultra-pure polypropylene (PP): Ensures chemical resistance and compliance with FDA 21 CFR 177.1520 & USP Class VI standards
  • High-clarity design: Crystal-clear walls for easy sample visibility
  • Secure O-ringed flat caps: ³/₄-turn design for leak-proof sealing and convenient opening/closing
  • Versatile temperature performance: Stable from −196 to +121 °C (not recommended for liquid nitrogen immersion)
  • Sterile and non-sterile options: Choose pre-sterilized assemblies or convenient non-sterile combo packs
  • Multiple formats: Available in the three most commonly used volumes, with conical or skirted/free-standing bottoms for different applications

VWR® screw cap microtubes provide reliable, contamination-free storage for your critical samples, reagents, and biological materials. Designed for durability and clarity, these high-quality tubes are trusted in life science, clinical, and research laboratories worldwide. Ideal for sample storage, molecular biology, PCR, centrifugation, and general lab use.

Certifications: Certified free of detectable DNase, RNase, DNA, and PCR inhibitors. Sterile items are also certified free of detectable endotoxins/pyrogens. Manufactured in compliance with stringent laboratory quality standards.

Specifications
  • Format:
    Conical bottom
  • Sterility:
    Non sterile
  • Volume:
    2.0 ml
  • Color:
    Clear
  • Description:
    Microtubes without ribbing
  • Packaging:
  • Maximum RCF:
    20000×g
  • Graduations:
    No
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.