$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
TYVEK 400 CUBRE 2XG CS25
85017-062
:
DUPONT SPECIALTY PRODUCTS USA, MX
TYVEK 400 CUBRE 2XG CS25
85017-062
:
DUPONT SPECIALTY PRODUCTS USA, MX
:
TY127S2X
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
: VWR International