$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
RETENER SPE CX CARTUCHO DE SILICONA 1 ML
85016-642
:
Thermo Scientific
RETENER SPE CX CARTUCHO DE SILICONA 1 ML
85016-642
:
Thermo Scientific
:
60107-301
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH