Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Registrarse
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RETENER PEP SPE CARTUCHO 1ML PSDVB/ UREA
Catalog # 85016-640
Supplier:
Thermo Scientific
Expand image
RETENER PEP SPE CARTUCHO 1ML PSDVB/ UREA
Catalog # 85016-640
Supplier:
Thermo Scientific
Supplier Number:
60107-201
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International