Skip to Header
Skip to Main Content
Skip to Footer
Avanzado
Avanzado
SDS
Certificados
Estructura química
Datos del fabricante
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MODULO DE PURIFICACION Q-GARD 1
Catalog # 85016-494
Proveedor:
MERCK, S.A. DE C.V. SV
Expand image
MODULO DE PURIFICACION Q-GARD 1
Catalog # 85016-494
Proveedor:
MERCK, S.A. DE C.V. SV
Supplier Number:
QGARD00R1
Frequently Bought Together
VWR® Disposable Serological Pipettes
Proveedor: VWR International
VWR® Nitrile Examination Gloves, Blue
Proveedor: VWR International
VWR® Disposable Petri Dishes
Proveedor: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Proveedor: BDH
VWR® Universal Pipette Tips
Proveedor: VWR International