Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Ingreso de pedido
Estado del pedido
Su cuenta
Carrito
Mexico
Ingreso de pedido
Estado del pedido
Su cuenta
Carrito
ContactUsLinkComponent
TETROXIDO DE OSMIO 1MG
Catalog # 85016-438
Supplier:
SIGMA ALDRICH QUIMICA,S DE RL MX
Expand image
TETROXIDO DE OSMIO 1MG
Catalog # 85016-438
Supplier:
SIGMA ALDRICH QUIMICA,S DE RL MX
Supplier Number:
419494-250MG
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
Supplier: VWR International