Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CRISOLES INCINERACION RONDA CUARZO 50ML
Catalog # 85015-502
Proveedor:
INSTRUMENTOS Y EQUIPOS FALCON MX
Ampliar imagen
CRISOLES INCINERACION RONDA CUARZO 50ML
Catalog # 85015-502
Proveedor:
INSTRUMENTOS Y EQUIPOS FALCON MX
Supplier Number:
303060
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Proveedor: VWR International
VWR® Universal Pipette Tips
Proveedor: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
Proveedor: VWR International
VWR® Nitrile Examination Gloves, Blue
Proveedor: VWR International
VWR® Disposable Petri Dishes
Proveedor: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Proveedor: BDH