Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CADA HYDRAVER 2 REACTIVO 100 PRUEBA MDB
Catalog # 85015-166
Supplier:
REACTIVOS Y EQUIP SA DE CV MX
Expand image
CADA HYDRAVER 2 REACTIVO 100 PRUEBA MDB
Catalog # 85015-166
Supplier:
REACTIVOS Y EQUIP SA DE CV MX
Supplier Number:
179032
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
Supplier: VWR International