Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Buchner Filtering Flasks

Socket Size 24/29, Side Socket Arm 0.90", 250 ml, 3.5×5"
Catalog # 470313-980
 :  CH0422C
Buchner Filtering Flasks
Catalog # 470313-980
Buchner Filtering Flasks
Buchner Filtering Flasks

Socket Size 24/29, Side Socket Arm 0.90", 250 ml, 3.5×5"

Price
$543.37
Log in for availability
Each (1UN)
Product Details & Documents

Buchner filtering flasks are made of high quality, heavy duty borosilicate glass.

 : Stopper not included.

Specifications
  • Capacity:
    250 ml
  • Description:
    Socket Size 24/29, Side Socket Arm 0.90"
  • Diameter ×H:
    3.5×5"