Order Entry
Mexico
Orders LinkContactUsLinkComponent
Volumetric Pipettes, Class A

Volumetric Pipettes, Class A

2 ml, Orange, ±0.010 ml
Catalog # 470312-908
Supplier #:  CH0559B
Volumetric Pipettes, Class A
Catalog # 470312-908
Volumetric Pipettes, Class A
Volumetric Pipettes, Class A

2 ml, Orange, ±0.010 ml

Price
$197.25
Log in for availability
Each (1UN)
Product Details & Documents

Reusable Class-A pipettes made from high quality borosilicate 3.3 glass.

  • Color coded for easy identification
  • Single graduation mark
  • Calibrated for delivery
  • Autoclavable

These Class-A volumetric pipettes are perfect for everyday laboratory use. They are designed for transferring liquid substances from one vessel to another with high accuracy.

Certifications: Calibrated according to DIN ISO 648 standards.

Specifications
  • Capacity:
    2 ml
  • Color Code:
    Orange
  • Tolerance:
    ±0.010 ml