Laboratory Aprons
Wide, 46×36"
470311-696
: LBAP04
Protective laboratory apron for multipurpose use.
- Flexible
- Chemical resistant
- Apron includes cloth ties at the neck and waist
These aprons are made of a PVC-coated fabric which stays flexible. There is reinforcement at the points of strain.
- Garment fit:Wide
- L×W:46×36"
- Cat. No.:470311-696
- Garment application:Chemical protection|General Purpose|Laboratory
- Gender:Unisex
- Fabric:PVC-coated fabric