Order Entry
Mexico
Orders LinkContactUsLinkComponent
Neoprene Stoppers
Neoprene Stoppers
5
Catalog # 470310-592
Supplier #:  CH0321H2H
Neoprene Stoppers
Catalog # 470310-592
Neoprene Stoppers
Neoprene Stoppers
5
Price
$513.78
Log in for availability
Paquete de 10
Product Details & Documents
Versatile neoprene stoppers perfect for every laboratory.
  • Solid, one hole or two hole stoppers are available
  • Manufacturered according to sizes recommended by ASTM standards
Manufactured from long lasting neoprene rubbers. Suitable for use with petroleum products, mineral oils, fats as well as most inorganic acids and bases. Neoprene rubber is oil resistant and suitable for temperatures up to 120 °C (250 °F).
Packaging: Packs of 10.
Specifications
  • Size:
    5