Order Entry
Mexico
Orders LinkContactUsLinkComponent
Estwing Chisel Head Hammers

Estwing Chisel Head Hammers

Chisel Hammer, 16 oz., 10.75"
Catalog # 470309-728
Supplier:   ESTWING MFG CO SE
Supplier #:  E3-16BLC
Estwing Chisel Head Hammers
Catalog # 470309-728
Supplier:  ESTWING MFG CO SE
Estwing Chisel Head Hammers
Estwing Chisel Head Hammers

Chisel Hammer, 16 oz., 10.75"

Price
$1,248.51
Log in for availability
Each (1UN)
Product Details & Documents

Ideal for use with sedimentary rocks.

  • Availabile in different head weights

Patented nylon end caps now makes these hammers the most durable on the market. The head and handles are forged in one piece and are fully polished. These tools have unsurpassed balance and are double tempered for added strength.

Specifications
  • Description:
    Chisel Hammer, 16 oz.
  • Length:
    10.75"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.