Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Estwing Chisel Head Hammers

Chisel Hammer, 20 oz., 11.25"
  470309-720
 :  E3-20BLC
Estwing Chisel Head Hammers
  470309-720
Estwing Chisel Head Hammers
Estwing Chisel Head Hammers

Chisel Hammer, 20 oz., 11.25"

 
$1,300.09
   
Each (1UN)
 

Ideal for use with sedimentary rocks.

  • Availabile in different head weights

Patented nylon end caps now makes these hammers the most durable on the market. The head and handles are forged in one piece and are fully polished. These tools have unsurpassed balance and are double tempered for added strength.

 
  • Description:
    Chisel Hammer, 20 oz.
  • Length:
    11.25"