Order Entry
Mexico
Orders LinkContactUsLinkComponent
Injection Training Pads

Injection Training Pads

Small Injection Pad "Paddy", 2.5", 2.5", 1.75"
Catalog # 470306-768
Supplier:   WALLCUR, LLC.
Supplier #:  REM-INJ-SP-01L
Injection Training Pads
Catalog # 470306-768
Supplier:  WALLCUR, LLC.
Injection Training Pads
Injection Training Pads

Small Injection Pad "Paddy", 2.5", 2.5", 1.75"

Price
$1,129.98
Log in for availability
Each (1UN)
Product Details & Documents

Remedy's medical training injection pads are designed for the practice of needle injections.

The pads can be used to simulate intradermal, intra-muscular and subcutaneous injections. The larger pad allows you to have more practice area, as well as practicing Z-track injections. A removable epidermis layer is placed on the surface of the pad allowing for the simulation of a "wheal" during intradermal injections.

The skin puncture resistance, as well as the resistance throughout the injection, closely mimics the feel of a real injection. Both pads have drainage on the bottom so they can be used with fluid filled syringes.

Ordering information: Additional epidermis layers may be purchased separately.

Specifications
  • Description:
    Small Injection Pad "Paddy"
  • Height:
    1.75"
  • Length:
    2.5"
  • Width:
    2.5"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.