Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
OVEROL MICROGARD 2000 STD MOD 122 XL
Catalog # 85013-296
Supplier:
ANSELL COMMERCIAL MEXICO SA DE MX
Expand image
OVEROL MICROGARD 2000 STD MOD 122 XL
Catalog # 85013-296
Supplier:
ANSELL COMMERCIAL MEXICO SA DE MX
Supplier Number:
WH20B3812205
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH