Order Entry
Mexico
Orders LinkContactUsLinkComponent
Tapes, Budnick Converting

Tapes, Budnick Converting

VHB Adhesive Transfer Tape, —, 2", 0.2"
Catalog # 76245-854
Supplier #:  F9469PCM200W2L
Tapes, Budnick Converting
Catalog # 76245-854
Tapes, Budnick Converting

VHB Adhesive Transfer Tape, —, 2", 0.2"

Price
$4,284.86
Log in for availability
Each (1UN)
Product Details & Documents

Documents

Specifications
  • Width:
    0.2"
  • Color:
  • Description:
    VHB Adhesive Transfer Tape
  • Length:
    2"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.