Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Ingreso de pedido
Estado del pedido
Su cuenta
Carrito
Mexico
Ingreso de pedido
Estado del pedido
Su cuenta
Carrito
ContactUsLinkComponent
TAPE SH SRS POLYESTER GREEN 0.5INX72 YD
Catalog # 76245-562
Supplier:
Caplugs
Expand image
TAPE SH SRS POLYESTER GREEN 0.5INX72 YD
Catalog # 76245-562
Supplier:
Caplugs
Supplier Number:
PC21-0500
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
Supplier: VWR International