Order Entry
Mexico
Orders LinkContactUsLinkComponent
Dispensing Tips, EFD
Dispensing Tips, EFD
Brush tip, standard soft bristle, 50.8 mm (2.00")
Catalog # 76242-298
Supplier #:  7018490
Dispensing Tips, EFD
Catalog # 76242-298
Dispensing Tips, EFD
Brush tip, standard soft bristle, 50.8 mm (2.00")
Price
$616.77
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
EFD produces the highest quality dispensing tips engineered hub flats for easy twist on, twist off. 360° SafetyLok™ thread ensures safe, positive attachment to syringe barrel.
  • Tapered Smooth flow for application of medium
  • Flexible Polypropylene shafts reach into hard-to-access areas
  • Free of flash, burrs and contaminants
  • Package labels include lot numbers for process control and traceability
For applications in chamfered, ESD-safe, PTFE-coated and PTFE-lined, microdot tips and oval tips.
Especificaciones
  • Description:
    Brush tip, standard soft bristle
  • Length:
    50.8 mm (2.00")