Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SOLDERING IRON F/MX-DSI DESOLDERING GUN
Catalog # 76241-494
Proveedor:
OK INTERNATIONAL TE
Expand image
SOLDERING IRON F/MX-DSI DESOLDERING GUN
Catalog # 76241-494
Proveedor:
OK INTERNATIONAL TE
Supplier Number:
MX-RM8E
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Proveedor: BDH
VWR® Disposable Serological Pipettes
Proveedor: VWR International
VWR® Disposable Petri Dishes
Proveedor: VWR International
VWR® Nitrile Examination Gloves, Blue
Proveedor: VWR International
VWR® Universal Pipette Tips
Proveedor: VWR International