Equal Volume Density Cube Sets
Density cube, set of 10
Catalog # 470303-936
Supplier: UNITED SCIENTIFIC SUPPLIES
: DCSET10
Jump to:
Product Details & Documents
High quality density cubes for the classroom.
- Ideal for density experiments and to explore the physical properties of different materials
- Compare masses for equal volumes
- Uniform, reliable dimensions
- Activity manual/reference guide included
Compare the masses and densities of these uniform 1" cubes. Explore the principles of bouyancy, and test the physical properties of metals and non-metals. Each set includes a reference table showing density values of individual specimens.
Ordering information: Set of 4 includes acrylic, pine, nylon, and aluminum, packaged in a poly bag.
Set of 10 includes acrylic, oak, pine, rosewood, steel, aluminum, copper, brass, nylon, and PVC, packaged in a wooden storage box.
Set of 12 includes acrylic, oak, pine, poplar, steel, aluminum, copper, brass, nylon, PVC, lignum vitae, and polypropylene, packaged in a plastic box.
Documents
Specifications
- Description:Density cube, set of 10



