$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
VOLUMETRIC FLASK WIDE MOUTH 30ML S. PE
85007-440
:
DWK LIFE SCIENCES S. DE R.L. DE MX
VOLUMETRIC FLASK WIDE MOUTH 30ML S. PE
85007-440
:
DWK LIFE SCIENCES S. DE R.L. DE MX
:
623015LP-30
Frequently Bought Together
VWR® Disposable Serological Pipettes
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Universal Pipette Tips
: VWR International