$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
LCR 0.45 UM PTFE 13 MM NON-STERILE
85005-532
:
MERCK, S.A. DE C.V. SV
LCR 0.45 UM PTFE 13 MM NON-STERILE
85005-532
:
MERCK, S.A. DE C.V. SV
:
SLCR013NL
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
: VWR International