$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
AUTOCLAVE HORIZONTAL MODELO DX-200
Catalog # 85005-232
Supplier:
LAB TECH INSTRUMENTACION SA DE MX
Expand image
AUTOCLAVE HORIZONTAL MODELO DX-200
Catalog # 85005-232
Supplier:
LAB TECH INSTRUMENTACION SA DE MX
Supplier Number:
2201-DX200
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH