Order Entry
Mexico
Orders LinkContactUsLinkComponent
Wire Triangles

Wire Triangles

Side length: 64 mm, 33 mm
Catalog # 470235-762
Supplier #:  4-2030-CS
Wire Triangles
Catalog # 470235-762
Wire Triangles
Wire Triangles

Side length: 64 mm, 33 mm

Price
$419.19
Log in for availability
Paquete de 10 Case de 10 (10UN)
Product Details & Documents

Perfect tool for suspending a crucible.

  • Uniform clay tubes
  • Galvanized iron wire resists rusting
Specifications
  • Ø:
    33 mm
  • Description:
    Side length: 64 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.