Order Entry
Mexico
Orders LinkContactUsLinkComponent
Wire Triangles

Wire Triangles

Side length: 38 mm, 20 mm
Catalog # 470235-758
Supplier #:  4-2019-CS
Wire Triangles
Catalog # 470235-758
Wire Triangles
Wire Triangles

Side length: 38 mm, 20 mm

Price
$295.59
Log in for availability
Paquete de 10 Case de 10 (10UN)
Product Details & Documents

Perfect tool for suspending a crucible.

  • Uniform clay tubes
  • Galvanized iron wire resists rusting
Specifications
  • Ø:
    20 mm
  • Description:
    Side length: 38 mm