Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
KF COULOMAT AK W/DIAFRAG HYDRANAL
Catalog # 85004-474
Supplier:
HONEYWELL INTERNATIONAL INC MX
Expand image
KF COULOMAT AK W/DIAFRAG HYDRANAL
Catalog # 85004-474
Supplier:
HONEYWELL INTERNATIONAL INC MX
Supplier Number:
34820-500ML-US
Frequently Bought Together
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
Supplier: VWR International