Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CEPILLO DE MANO PARA TALLAR GRIS 210X65M
Catalog # 85013-210
Supplier:
FBK MEXICO, S.A. DE C.V. MX
Expand image
CEPILLO DE MANO PARA TALLAR GRIS 210X65M
Catalog # 85013-210
Supplier:
FBK MEXICO, S.A. DE C.V. MX
Supplier Number:
54153-11
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International